Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ext1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130540.1 | NP_001124012.1 | 7 - 8 | 1751 | 68 | |
GenBank mRNA | BC105839.1 | - | 7 - 8 | 1066 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |