Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kiss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181692.1 | NP_859043.1 | 1 - 2 | 107 | 65 | |
XM_008769443.1 | XP_008767665.1 | 2 - 3 | 560 | 65 | ||
GenBank mRNA | AY196983.1 | - | 1 - 2 | 107 | 65 | |
JX139030.1 | - | 2 - 3 | 518 | 65 | ||
JX139031.1 | - | 2 - 3 | 314 | 65 | ||
JX139032.1 | - | 2 - 3 | 239 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons |