Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182814.2 | NP_877966.1 | 8 - 9 | 1052 | 136 | |
GenBank mRNA | AY223861.1 | - | 8 - 9 | 918 | 136 | |
BC079283.1 | - | 8 - 9 | 1023 | 7134210 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |