Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017243.2 | NP_058939.1 | 5 - 6 | 815 | 100 | |
GenBank mRNA | BC078853.1 | - | 5 - 6 | 813 | 7190069 | 100 |
M17258.1 | - | 5 - 6 | 791 | 100 | ||
M29392.1 | - | 5 - 6 | 791 | 100 | ||
X16554.1 | - | 5 - 6 | 812 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |