Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109469.1 | NP_001102939.1 | 1 - 2 | 402 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |