Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dedd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748789.2 | XP_003748837.1 | 1 - 2 | 122 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Rattus norvegicus Probe spans exons |