Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010956.2 | NP_001010956.1 | 8 - 9 | 1145 | 105 | |
NM_001033931.1 | NP_001029103.1 | 8 - 9 | 1145 | 105 | ||
XM_006236832.2 | XP_006236894.1 | 7 - 8 | 1094 | 105 | ||
GenBank mRNA | AY862307.1 | - | 8 - 9 | 972 | 105 | |
AY862308.1 | - | 8 - 9 | 940 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |