Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012473.1 | NP_001012491.1 | 3 - 4 | 232 | 81 | |
GenBank mRNA | BC089905.1 | - | 3 - 4 | 232 | 7322752 | 81 |
FQ218345.1 | - | 3 - 4 | 149 | 81 | ||
FQ226282.1 | - | 3 - 4 | 271 | 81 | ||
FQ227427.1 | - | 3 - 4 | 252 | 81 | ||
FQ235323.1 | - | 3 - 4 | 271 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |