Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017348.2 | NP_059044.1 | 2 - 3 | 983 | 67 | |
XM_006229611.1 | XP_006229673.1 | 2 - 3 | 1015 | 67 | ||
GenBank mRNA | BC168238.1 | - | 2 - 3 | 961 | 5598758 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |