Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053656.2 | NP_446108.2 | 2 - 3 | 309 | 67 | |
XM_006249515.2 | XP_006249577.1 | 2 - 3 | 502 | 67 | ||
XM_006249516.2 | XP_006249578.1 | 2 - 3 | 501 | 67 | ||
XM_006249517.2 | XP_006249579.1 | 2 - 3 | 464 | 67 | ||
XM_008769305.1 | XP_008767527.1 | 2 - 3 | 466 | 67 | ||
XM_008769306.1 | XP_008767528.1 | 2 - 3 | 178 | 67 | ||
GenBank mRNA | AF020756.1 | - | 2 - 3 | 290 | 67 | |
AF020757.1 | - | 2 - 3 | 336 | 67 | ||
AF020758.1 | - | 1 - 2 | 430 | 67 | ||
AF020759.1 | - | 2 - 3 | 271 | 67 | ||
U14414.1 | - | 2 - 3 | 309 | 67 | ||
Y10473.1 | - | 2 - 3 | 309 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |