Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039035.2 | NP_001034124.2 | 7 - 8 | 756 | 73 | |
XM_006239642.1 | XP_006239704.1 | 7 - 8 | 787 | 73 | ||
XM_006239643.1 | XP_006239705.1 | 7 - 8 | 763 | 73 | ||
GenBank mRNA | FM079490.1 | - | 2 - 3 | 153 | 73 | |
FM092264.1 | - | 4 - 5 | 231 | 73 | ||
FQ229134.1 | - | 6 - 7 | 1613 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |