Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ajap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101014.1 | NP_001094484.1 | 4 - 5 | 1449 | 110 | |
GenBank mRNA | AB179740.1 | - | 4 - 5 | 1449 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |