Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163277.1 | NP_001156749.1 | 2 - 3 | 743 | 87 | |
XM_008760004.1 | XP_008758226.1 | 6 - 7 | 1838 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |