Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239171.2 | XP_006239233.1 | 9 - 10 | 1865 | 100 | |
XM_006239174.2 | XP_006239236.1 | 8 - 9 | 1792 | 100 | ||
XM_006239177.2 | XP_006239239.1 | 8 - 9 | 842 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |