Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106131.1 | NP_001099601.1 | 7 - 8 | 1174 | 75 | |
XM_006254942.2 | XP_006255004.1 | 7 - 8 | 2279 | 75 | ||
XM_006254943.2 | XP_006255005.1 | 7 - 8 | 2280 | 75 | ||
XM_006254944.2 | XP_006255006.1 | 7 - 8 | 2281 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Rattus norvegicus Probe spans exons |