Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022665.3 | NP_073156.3 | 5 - 6 | 701 | 107 | |
XM_008767194.1 | XP_008765416.1 | 6 - 7 | 736 | 107 | ||
GenBank mRNA | X17611.1 | - | 5 - 6 | 700 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |