Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022622.1 | NP_072144.1 | 2 - 3 | 207 | 69 | |
GenBank mRNA | AF182946.1 | - | 2 - 3 | 207 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |