Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duxbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252678.2 | XP_006252740.1 | 2 - 3 | 3087 | 62 | |
XM_006252679.2 | XP_006252741.1 | 3 - 4 | 1088 | 62 | ||
XM_008771031.1 | XP_008769253.1 | 1 - 2 | 3762 | 62 | ||
XM_224643.6 | XP_224643.1 | 2 - 3 | 1763 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |