Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013909.1 | NP_001013931.1 | 4 - 5 | 314 | 63 | |
GenBank mRNA | BC089903.1 | - | 4 - 5 | 314 | 7384104 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |