Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012606.2 | NP_036738.1 | 4 - 5 | 551 | 68 | |
XM_006243917.2 | XP_006243979.2 | 4 - 5 | 652 | 68 | ||
GenBank mRNA | BC081832.1 | - | 4 - 5 | 551 | 7192037 | 68 |
FQ216106.1 | - | 4 - 5 | 563 | 68 | ||
FQ216251.1 | - | 4 - 5 | 563 | 68 | ||
FQ216255.1 | - | 4 - 5 | 583 | 68 | ||
FQ217222.1 | - | 4 - 5 | 566 | 68 | ||
FQ223547.1 | - | 4 - 5 | 563 | 68 | ||
X14812.1 | - | 4 - 5 | 544 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |