Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105868.1 | NP_001099338.1 | 14 - 15 | 3128 | 92 | |
XM_006248561.2 | XP_006248623.1 | 14 - 15 | 3256 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |