Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107078.2 | NP_001100548.1 | 3 - 4 | 1672 | 68 | |
XM_006248693.2 | XP_006248755.1 | 4 - 5 | 2045 | 68 | ||
XM_006248694.2 | XP_006248756.1 | 4 - 5 | 1865 | 68 | ||
XM_006248695.2 | XP_006248757.1 | 4 - 5 | 2042 | 68 | ||
XM_006248696.2 | XP_006248758.1 | 3 - 4 | 1900 | 68 | ||
XM_006248697.2 | XP_006248759.1 | 3 - 4 | 1897 | 68 | ||
XM_006248698.2 | XP_006248760.1 | 3 - 4 | 1749 | 68 | ||
GenBank mRNA | BC169079.1 | - | 3 - 4 | 1672 | 7937326 | 68 |
FQ214252.1 | - | 3 - 4 | 1866 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |