Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182736.1 | NP_877400.1 | 6 - 7 | 563 | 61 | |
XM_008768795.1 | XP_008767017.1 | 7 - 8 | 1328 | 61 | ||
GenBank mRNA | AJ534984.1 | - | 6 - 7 | 563 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |