Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199486.2 | NP_955780.1 | 1 | 1088 | 67 | |
GenBank mRNA | AB113394.1 | - | 1 | 1088 | 67 | |
AY573973.1 | - | 1 | 1088 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |