Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024240.1 | NP_001019411.1 | 3 - 4 | 406 | 80 | |
GenBank mRNA | BC093397.1 | - | 3 - 4 | 406 | 7389084 | 80 |
FQ234743.1 | - | 3 - 4 | 440 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |