Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199376.1 | NP_955408.1 | 6 - 7 | 743 | 83 | |
XM_006254542.2 | XP_006254604.1 | 6 - 7 | 777 | 83 | ||
XM_008772024.1 | XP_008770246.1 | 6 - 7 | 1065 | 83 | ||
XM_008772025.1 | XP_008770247.1 | 6 - 7 | 829 | 83 | ||
GenBank mRNA | BC062050.1 | - | 6 - 7 | 743 | 5598825 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |