Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019149.3 | NP_062022.2 | 8 - 9 | 1671 | 99 | |
XM_006254534.2 | XP_006254596.1 | 8 - 9 | 1693 | 99 | ||
XM_006254535.2 | XP_006254597.1 | 8 - 9 | 1644 | 99 | ||
GenBank mRNA | AB205483.1 | - | 7 - 8 | 2441 | 99 | |
M63485.1 | - | 8 - 9 | 1656 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |