Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019149.3 | NP_062022.2 | 9 - 10 | 1834 | 90 | |
XM_006254534.2 | XP_006254596.1 | 9 - 10 | 1856 | 90 | ||
XM_006254535.2 | XP_006254597.1 | 9 - 10 | 1807 | 90 | ||
GenBank mRNA | AB205483.1 | - | 8 - 9 | 2604 | 90 | |
M63485.1 | - | 9 - 10 | 1819 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |