Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108750.1 | NP_001102220.1 | 15 - 16 | 1311 | 77 | |
GenBank mRNA | AM747284.1 | - | 15 - 16 | 1182 | 77 | |
BC168840.1 | - | 15 - 16 | 1234 | 7125016 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |