Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242180.2 | XP_006242242.1 | 15 - 16 | 2369 | 63 | |
GenBank mRNA | BC085761.1 | - | 15 - 16 | 1925 | 7193798 | 63 |
D43623.1 | - | 14 - 15 | 1900 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |