Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242180.2 | XP_006242242.1 | 8 - 9 | 1374 | 89 | |
GenBank mRNA | BC085761.1 | - | 8 - 9 | 930 | 7193798 | 89 |
D43623.1 | - | 7 - 8 | 905 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |