Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170557.1 | NP_001164028.1 | 14 - 15 | 1698 | 88 | |
XM_006240921.2 | XP_006240983.1 | 14 - 15 | 1709 | 88 | ||
XM_006240922.2 | XP_006240984.1 | 14 - 15 | 1652 | 88 | ||
XM_006240923.2 | XP_006240985.1 | 13 - 14 | 1592 | 88 | ||
XM_008765099.1 | XP_008763321.1 | 15 - 16 | 1681 | 88 | ||
XM_008765100.1 | XP_008763322.1 | 14 - 15 | 1839 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase 3' Most Rattus norvegicus 1700 Probe spans exons |