Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241090.1 | XP_006241152.1 | 3 - 4 | 431 | 137 | |
XM_006241091.2 | XP_006241153.1 | 3 - 4 | 540 | 137 | ||
XM_006241092.1 | XP_006241154.1 | 2 - 3 | 343 | 137 | ||
XM_008765183.1 | XP_008763405.1 | 2 - 3 | 771 | 137 | ||
XM_234837.7 | XP_234837.3 | 3 - 4 | 872 | 137 | ||
GenBank mRNA | FQ210490.1 | - | 3 - 4 | 436 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |