Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 3 - 4 | 176 | 82 | |
NM_022516.5 | NP_071961.2 | 3 - 4 | 176 | 82 | ||
GenBank mRNA | BC061858.1 | - | 3 - 4 | 171 | 5598495 | 82 |
CB772933.1 | - | 3 - 4 | 186 | 82 | ||
FN803753.1 | - | 3 - 4 | 146 | 82 | ||
X60789.1 | - | 3 - 4 | 139 | 82 | ||
X74565.1 | - | 3 - 4 | 157 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |