Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107170.1 | NP_001100640.1 | 1 - 2 | 149 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |