Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271109.1 | NP_001258038.1 | 10 - 11 | 1341 | 83 | |
NM_001271110.1 | NP_001258039.1 | 10 - 11 | 1369 | 83 | ||
NM_080479.2 | NP_536727.2 | 10 - 11 | 1341 | 83 | ||
XM_006237924.2 | XP_006237986.1 | 10 - 11 | 1431 | 83 | ||
GenBank mRNA | AJ293617.1 | - | 9 - 10 | 1282 | 83 | |
AY724507.1 | - | 10 - 11 | 1354 | 83 | ||
BC086998.1 | - | 8 - 9 | 2298 | 83 | ||
BC107467.1 | - | 10 - 11 | 1341 | 7459969 | 83 | |
FQ229095.1 | - | 10 - 11 | 1354 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |