Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271109.1 | NP_001258038.1 | 7 - 8 | 1146 | 64 | |
NM_001271110.1 | NP_001258039.1 | 7 - 8 | 1174 | 64 | ||
NM_080479.2 | NP_536727.2 | 7 - 8 | 1146 | 64 | ||
XM_006237924.2 | XP_006237986.1 | 7 - 8 | 1236 | 64 | ||
GenBank mRNA | AJ293617.1 | - | 6 - 7 | 1087 | 64 | |
AY724507.1 | - | 7 - 8 | 1159 | 64 | ||
BC086998.1 | - | 6 - 7 | 1777 | 64 | ||
BC107467.1 | - | 7 - 8 | 1146 | 7459969 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |