Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227719.2 | XP_006227781.1 | 1 - 2 | 447 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |