Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053423.1 | NP_445875.1 | 13 - 14 | 3009 | 86 | |
XM_008758683.1 | XP_008756905.1 | 13 - 14 | 4256 | 86 | ||
GenBank mRNA | AY539717.1 | - | 13 - 14 | 3009 | 86 | |
AY539718.1 | - | 13 - 14 | 2991 | 86 | ||
AY539719.1 | - | 14 - 15 | 3114 | 86 | ||
AY539720.1 | - | 13 - 14 | 3082 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |