Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248842.2 | XP_006248904.1 | 10 - 11 | 1930 | 142 | |
XM_006248844.2 | XP_006248906.1 | 10 - 11 | 1930 | 142 | ||
XM_006248845.2 | XP_006248907.1 | 10 - 11 | 1933 | 142 | ||
XM_346914.7 | XP_346915.4 | 9 - 10 | 1767 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |