Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762677.1 | XP_008760899.1 | 3 - 4 | 1315 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |