Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019128.4 | NP_062001.1 | 1 - 2 | 2353 | 87 | |
GenBank mRNA | X52017.1 | - | 1 - 2 | 1080 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |