Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108527.1 | NP_001101997.1 | 2 - 3 | 201 | 98 | |
XM_006231684.2 | XP_006231746.1 | 1 - 2 | 2262 | 98 | ||
GenBank mRNA | FQ215895.1 | - | 2 - 3 | 190 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |