Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077674.1 | NP_001071142.1 | 3 - 4 | 1270 | 89 | |
GenBank mRNA | BC127513.1 | - | 3 - 4 | 1270 | 7099599 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |