Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105985.1 | NP_001099455.1 | 3 - 4 | 1137 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |