Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001067807.5 | XP_001067807.3 | 13 - 14 | 1710 | 172 | |
XM_006250417.1 | XP_006250479.1 | 14 - 15 | 1549 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |