Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001067807.5 | XP_001067807.3 | 14 - 15 | 1809 | 132 | |
XM_006250417.1 | XP_006250479.1 | 15 - 16 | 1648 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |