Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_223012.8 | XP_223012.6 | 6 - 7 | 890 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |