Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057098.1 | NP_476439.1 | 9 - 10 | 973 | 60 | |
XM_006235740.1 | XP_006235802.1 | 9 - 10 | 1065 | 60 | ||
XM_006235741.1 | XP_006235803.1 | 9 - 10 | 1050 | 60 | ||
XM_006235742.2 | XP_006235804.1 | 9 - 10 | 1039 | 60 | ||
GenBank mRNA | D12927.1 | - | 9 - 10 | 973 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |