Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106201.1 | NP_001099671.1 | 2 - 3 | 2214 | 84 | |
XM_006231634.1 | XP_006231696.1 | 3 - 4 | 2890 | 84 | ||
XM_006231635.2 | XP_006231697.1 | 3 - 4 | 2828 | 84 | ||
XM_008760523.1 | XP_008758745.1 | 3 - 4 | 2890 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |